Merck
  • Metabolomic analysis uncovered an association of serum phospholipid levels with estrogen-induced mammary tumors in female ACI/Seg rats.
Metabolomic analysis uncovered an association of serum phospholipid levels with estrogen-induced mammary tumors in female ACI/Seg rats.
Toxicology letters (2018-02-20)
Yoshinori Okamoto, Akira Aoki, Koji Ueda, Hideto Jinno
ABSTRACT
Estrogen is reported to be involved in mammary tumorigenesis. To unveil metabolic signatures for estrogen-induced mammary tumorigenesis, we carried out serum metabolomic analysis in an estrogen-induced mammary tumor model, female August Copenhagen-Irish/Segaloff (ACI/Seg) rats, using liquid chromatography-mass spectrometry. In contrast to the control group, all rats with an implanted 17β-estradiol (E2) pellet developed mammary tumors during this experiment. E2 treatment significantly suppressed body weight gain. But no significant differences in food consumption were observed between the two groups, suggesting that metabolic alteration depended on E2 treatment. Serum metabolomic analysis detected 116 features that were statistically different (p < 0.01) between the groups. Quantitation analysis revealed that several phospholipids such as phosphatidylcholines and lysophosphatidylcholines (LPCs) were identified as significantly different metabolites. E2-treated rat serum stimulated the proliferation of human breast cancer MDA-MB-231 cells. In addition, the proliferation effect was diminished by pretreating cells with either autotaxin inhibitor or antagonist for lysophosphatidic acid receptor whose ligands are metabolites of LPCs via autotaxin-mediated hydrolysis. In summary, our results suggest that not only are phospholipids potential biomarkers for mammary tumors but importantly, LPCs themselves could be associated with E2-induced mammary tumorigenesis in female ACI/Seg rats.
Sigma-Aldrich
S32826 disodium salt hydrate, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Oleoyl-sn-glycero-3-phosphocholine, synthetic, ≥99%