MilliporeSigma
  • Apoptotic cell-induced AhR activity is required for immunological tolerance and suppression of systemic lupus erythematosus in mice and humans.
Apoptotic cell-induced AhR activity is required for immunological tolerance and suppression of systemic lupus erythematosus in mice and humans.
Nature immunology (2018-05-16)
Rahul Shinde, Kebria Hezaveh, Marie Jo Halaby, Andreas Kloetgen, Ankur Chakravarthy, Tiago da Silva Medina, Reema Deol, Kieran P Manion, Yuriy Baglaenko, Maria Eldh, Sara Lamorte, Drew Wallace, Sathi Babu Chodisetti, Buvana Ravishankar, Haiyun Liu, Kapil Chaudhary, David H Munn, Aristotelis Tsirigos, Michael Madaio, Susanne Gabrielsson, Zahi Touma, Joan Wither, Daniel D De Carvalho, Tracy L McGaha
ABSTRACT
The transcription factor AhR modulates immunity at multiple levels. Here we report that phagocytes exposed to apoptotic cells exhibited rapid activation of AhR, which drove production of the cytokine IL-10. Activation of AhR was dependent on interactions between apoptotic-cell DNA and the pattern-recognition receptor TLR9 that was required for the prevention of immune responses to DNA and histones in vivo. Moreover, disease progression in mouse systemic lupus erythematosus (SLE) correlated with strength of the AhR signal, and the disease course could be altered by modulation of AhR activity. Deletion of AhR in the myeloid lineage caused systemic autoimmunity in mice, and an enhanced AhR transcriptional signature correlated with disease in patients with SLE. Thus, AhR activity induced by apoptotic cell phagocytes maintains peripheral tolerance.
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)