MilliporeSigma
  • Endothelial barrier function is differentially regulated by CEACAM1-mediated signaling.
Endothelial barrier function is differentially regulated by CEACAM1-mediated signaling.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-05-11)
Sharang Ghavampour, Florian Kleefeldt, Heike Bömmel, Julian Volland, Alexander Paus, Andrea Horst, Verena Pfeiffer, Stefan Hübner, Nicole Wagner, Uwe Rueckschloss, Süleyman Ergün
ABSTRACT
Carcinoembryonic antigen-related cell adhesion molecule-1 (CEACAM1) is known to be crucial to vasculogenesis and angiogenesis. Recently, CEACAM1 deficiency was shown to result in the formation of aortic plaque-like lesions, indicating a role for CEACAM1 in adult vessels as well. The underlying mechanisms remained largely elusive. Therefore, we aimed to elucidate the role of CEACAM1 in endothelial homeostasis. Here, we show that CEACAM1 deficiency causes subcellular eNOS redistribution in endothelial cells ( i.e., by eNOS depalmitoylation) and alters endothelial glycocalyx that confers antiadhesive properties to the endothelium ( i.e., by repression of glycocalyx-degrading enzymes). Accordingly, our analysis revealed an increased leukocyte-endothelial interaction in CEACAM1-deficient endothelium. In addition, CEACAM1 age dependently modulated basal and TNF-α-mediated endothelial barrier (EB) leakiness. In younger mice, CEACAM1 was protective for EB, whereas in aged mice it promoted EB leakiness. EB function depends on interendothelial adherence junctions formed by β-catenin/vascular endothelial-cadherin complexes. We show here that CEACAM1 influenced basal and TNF-α-mediated phosphorylation of β-catenin and caveolin-1, which are essential players in EB modulation. Both increased adhesiveness to leukocytes and EB modulation due to CEACAM1 deficiency may facilitate inflammatory cell transmigration into the vascular wall and subsequent plaque formation. Collectively, these results identify a crucial role for CEACAM1 in endothelial homeostasis of adult blood vessels.-Ghavampour, S., Kleefeldt, F., Bömmel, H., Volland, J., Paus, A., Horst, A., Pfeiffer, V., Hübner, S., Wagner, N., Rueckschloss, U., Ergün, S. Endothelial barrier function is differentially regulated by CEACAM1-mediated signaling.
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)