MilliporeSigma
  • Acetoacetanilides as masked isocyanates: facile and efficient synthesis of unsymmetrically substituted ureas.
Acetoacetanilides as masked isocyanates: facile and efficient synthesis of unsymmetrically substituted ureas.
Organic letters (2010-09-03)
Ying Wei, Jing Liu, Shaoxia Lin, Hongqian Ding, Fushun Liang, Baozhong Zhao
ABSTRACT
A general and practical method for the preparation of unsymmetrically substituted ureas has been developed. By the reactions of acetoacetanilides with various amines including primary/secondary amines, a series of substituted aryl ureas were achieved in high yields. Acetoacetanilide substrates can be considered as masked reagents that liberate reactive isocyanates in situ.
Sigma-Aldrich
2-Methylpiperidine, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Sigma-Aldrich
Acetoacetanilide, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20