Merck
The Atypical Chemokine Receptor ACKR2 is Protective Against Sepsis.
Shock (Augusta, Ga.) (2018-03-29)
Fernanda V E Silva Castanheira, Vanessa Borges, Fabiane Sônego, Alexandre Kanashiro, Paula B Donate, Paulo H Melo, Kenneth Pallas, Remo C Russo, Flávio A Amaral, Mauro M Teixeira, Fernando S Ramalho, Thiago M Cunha, Foo Y Liew, José C Alves-Filho, Gerard J Graham, Fernando Q Cunha
ABSTRACT
Sepsis is a systemic inflammatory response as a result of uncontrolled infections. Neutrophils are the first cells to reach the primary sites of infection, and chemokines play a key role in recruiting neutrophils. However, in sepsis chemokines could also contribute to neutrophil infiltration to vital organs leading to multiple organ failure. ACKR2 is an atypical chemokine receptor, which can remove and degrade inflammatory CC chemokines. The role of ACK2 in sepsis is unknown. Using a model of cecal ligation and puncture (CLP), we demonstrate here that ACKR2 deficient () mice exhibited a significant reduction in the survival rate compared with similarly treated wild-type (WT) mice. However, neutrophil migration to the peritoneal cavity and bacterial load were similar between WT and ACKR2 mice during CLP. In contrast, ACKR2 mice showed increased neutrophil infiltration and elevated CC chemokine levels in the lung, kidney, and heart compared with the WT mice. In addition, ACKR2 mice also showed more severe lesions in the lung and kidney than those in the WT mice. Consistent with these results, WT mice under nonsevere sepsis (90% survival) had higher expression of ACKR2 in these organs than mice under severe sepsis (no survival). Finally, the lungs from septic patients showed increased number of ACKR2 cells compared with those of nonseptic patients. Our data indicate that ACKR2 may have a protective role during sepsis, and the absence of ACKR2 leads to exacerbated chemokine accumulation, neutrophil infiltration, and damage to vital organs.
Sigma-Aldrich
Ertapenem sodium, ≥90% (HPLC)