MilliporeSigma
Tubulin structure and biochemistry.
Current opinion in cell biology (1992-02-01)
R F Ludueña, A Banerjee, I A Khan
ABSTRACT
In the past year, much has been learned about structure-function correlations in the tubulin molecule, and specifically about the nature and roles of post-translational modifications and tubulin isotypes. The interactions between tubulin and its ligands--both microtubule-associated proteins and anti-mitotic drugs--are becoming clearer at the molecular level.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mg
In Stock
Details...
$538.00
$376.60
10 mg
In Stock
Details...
$1,840.00