MilliporeSigma
  • Tuneable poration: host defense peptides as sequence probes for antimicrobial mechanisms.
Tuneable poration: host defense peptides as sequence probes for antimicrobial mechanisms.
Scientific reports (2018-10-10)
Marc-Philipp Pfeil, Alice L B Pyne, Valeria Losasso, Jascindra Ravi, Baptiste Lamarre, Nilofar Faruqui, Hasan Alkassem, Katharine Hammond, Peter J Judge, Martyn Winn, Glenn J Martyna, Jason Crain, Anthony Watts, Bart W Hoogenboom, Maxim G Ryadnov
ABSTRACT
The spread of antimicrobial resistance stimulates discovery strategies that place emphasis on mechanisms circumventing the drawbacks of traditional antibiotics and on agents that hit multiple targets. Host defense peptides (HDPs) are promising candidates in this regard. Here we demonstrate that a given HDP sequence intrinsically encodes for tuneable mechanisms of membrane disruption. Using an archetypal HDP (cecropin B) we show that subtle structural alterations convert antimicrobial mechanisms from native carpet-like scenarios to poration and non-porating membrane exfoliation. Such distinct mechanisms, studied using low- and high-resolution spectroscopy, nanoscale imaging and molecular dynamics simulations, all maintain strong antimicrobial effects, albeit with diminished activity against pathogens resistant to HDPs. The strategy offers an effective search paradigm for the sequence probing of discrete antimicrobial mechanisms within a single HDP.
Sigma-Aldrich
HBTU, ≥98.0% (T)