MilliporeSigma
  • First study on trichothecene and zearalenone exposure of the Romanian population through wheat-based products consumption.
First study on trichothecene and zearalenone exposure of the Romanian population through wheat-based products consumption.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2018-09-15)
Oana Stanciu, Cristina Juan, Doina Miere, Houda Berrada, Felicia Loghin, Jordi Mañes
ABSTRACT
In this study, a dietary exposure assessment of mycotoxins was conducted for the Romanian population using the contamination data of a various categories of wheat-based products for direct human consumption. Wheat-based foods (n = 181) commercialized in Romania, including flour, bread, biscuits, breakfast cereals and pasta, were evaluated by GC-QqQ-MS/MS for the occurrence of deoxynivalenol (DON), 3-acetyldeoxynivalenol (3AcDON), 15-acetyldeoxynivalenol (15AcDON), fusarenon-X, nivalenol, HT-2 and T-2 toxins, diacetoxyscirpenol, neosolaniol and zearalenone (ZEA). DON and 15AcDON were detected in 63 and 5% of all the analyzed samples, whereas 13-AcDON, HT-2, T-2, NIV and ZEA were not detected. Exposure of Romanian adult population was assessed, the EDIs for the sum of DON+3AcDON+15AcDON were 669 ng kg-1 bw day-1 at low-bound estimation, and 690 ng kg-1 bw day-1 at upper-bound estimation, being lower than the TDI set (1000 ng kg-1 bw day-1).
Product Description
Supelco
Neosolaniol