MilliporeSigma
  • ERKs and p38 kinases mediate ultraviolet B-induced phosphorylation of histone H3 at serine 10.
ERKs and p38 kinases mediate ultraviolet B-induced phosphorylation of histone H3 at serine 10.
The Journal of biological chemistry (2000-05-12)
S P Zhong, W Y Ma, Z Dong
ABSTRACT
Histone H3 is the core protein of the nucleosome. Phosphorylation of H3 involves immediate early gene expression, chromatin remodeling, and chromosome condensation during mitosis. Very recently, Rsk2 or MSK1 kinase-mediated phosphorylation of H3 at serine 10 was reported. In the present study, we show that both ERKs and p38 kinase may mediate ultraviolet B-induced phosphorylation of H3 at serine 10. PD 98059, a MEK1 inhibitor, and SB 202190, a p38 kinase inhibitor, efficiently inhibited ultraviolet B-induced phosphorylation of H3. Phosphorylation of H3 was also inhibited in cells expressing dominant negative mutant (DNM) ERK2 and DNM p38 kinase. In contrast, no inhibition of H3 phosphorylation in Jnk1 or Jnk2 knockout cells (Jnk1(-/-) or Jnk2(-/-)) and cells expressing DNM JNK1 was observed. More importantly, incubation of active ERK2 or p38 kinase with H3 protein resulted in phosphorylation of H3 at serine 10 in vitro. These results suggest that ERK and p38 kinase are at least two important mediators of phosphorylation of H3 at serine 10.
Sigma-Aldrich
JNK2α2/SAPK1a Protein, active, 10 µg, Active, N-terminal His-tagged, full length human JNK2α2/SAPK1a, for use in Kinase Assays.
Sigma-Aldrich
MAP Kinase 2/Erk2 Protein, active, mouse, 10 µg, Active, N-terminal GST-tagged, recombinant MAP Kinase 2, expressed in E. coli cells, activated using MEK1 (Catalog #14-429). For use in Kinase Assays.