MilliporeSigma
  • Recent molecular approaches to elucidate the mechanism of embryo implantation: trophinin, bystin, and tastin as molecules involved in the initial attachment of blastocysts to the uterus in humans.
Recent molecular approaches to elucidate the mechanism of embryo implantation: trophinin, bystin, and tastin as molecules involved in the initial attachment of blastocysts to the uterus in humans.
Seminars in reproductive medicine (2001-04-13)
R Aoki, M N Fukuda
ABSTRACT
Elucidation of the implantation mechanism in humans at the molecular level has been difficult because of methodological restrictions. Instead of using human materials during the implantation period, two human tumor cell lines that respectively mimic the biological behaviors of a blastocyst and uterine luminal epithelial cells were utilized successfully to identify three novel adhesion molecules named trophinin, bystin, and tastin. Trophinin is a membrane protein strongly expressed both on the apical surface of the trophectoderm of a simian blastocyst and at a putative implantation site of the human endometrium. Bystin and tastin are cytoplasmic proteins that associate with trophinin by presumably forming an active adhesion machinery. The expression patterns of these molecules are suggestive of their involvement in the initial blastocyst attachment to the uterus as well as in the subsequent placental development. Future perspectives in molecular implantation research are also discussed in relation to breakthroughs in assisted reproduction.