- A new synthesis of pyrrolidines by way of an enantioselective Mannich/diastereoselective hydroamination reaction sequence.
A new synthesis of pyrrolidines by way of an enantioselective Mannich/diastereoselective hydroamination reaction sequence.
Organic letters (2011-07-14)
Jenny M Baxter Vu, James L Leighton
PMID21749067
ABSTRACT
A new two-step synthesis of highly substituted pyrrolidines has been developed. Chiral silane Lewis acid promoted enantioselective Mannich reactions of silyl ketene imines with acylhydrazones may be used to access bishomoallylic benzoic hydrazides that in turn may be cyclized to pyrrolidines by way of the thermal hydroamination reaction reported recently by Beauchemin. Importantly, excellent diastereoselectivity may be realized in the hydroamination reactions.