Merck
  • Ordered mesoporous silica (OMS) as an adsorbent and membrane for separation of carbon dioxide (CO2).
Ordered mesoporous silica (OMS) as an adsorbent and membrane for separation of carbon dioxide (CO2).
Advances in colloid and interface science (2010-01-12)
Thiam-Leng Chew, Abdul L Ahmad, Subhash Bhatia
ABSTRACT
Separation of carbon dioxide (CO(2)) from gaseous mixture is an important issue for the removal of CO(2) in natural gas processing and power plants. The ordered mesoporous silicas (OMS) with uniform pore structure and high density of silanol groups, have attracted the interest of researchers for separation of carbon dioxide (CO(2)) using adsorption process. These mesoporous silicas after functionalization with amino groups have been studied for the removal of CO(2). The potential of functionalized ordered mesoporous silica membrane for separation of CO(2) is also recognized. The present paper reviews the synthesis of mesoporous silicas and important issues related to the development of mesoporous silicas. Recent studies on the CO(2) separation using ordered mesoporous silicas (OMS) as adsorbent and membrane are highlighted. The future prospectives of mesoporous silica membrane for CO(2) adsorption and separation are also presented and discussed.
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Pricing and availability is not currently available.