MilliporeSigma
  • L-allo-threonine aldolase from Aeromonas jandaei DK-39: gene cloning, nucleotide sequencing, and identification of the pyridoxal 5'-phosphate-binding lysine residue by site-directed mutagenesis.
L-allo-threonine aldolase from Aeromonas jandaei DK-39: gene cloning, nucleotide sequencing, and identification of the pyridoxal 5'-phosphate-binding lysine residue by site-directed mutagenesis.
Journal of bacteriology (1997-06-01)
J Q Liu, T Dairi, M Kataoka, S Shimizu, H Yamada
ABSTRACT
We have isolated the gene encoding L-allo-threonine aldolase (L-allo-TA) from Aeromonas jandaei DK-39, a pyridoxal 5'-phosphate (PLP)-dependent enzyme that stereospecifically catalyzes the interconversion of L-allo-threonine and glycine. The gene contains an open reading frame consisting of 1,014 nucleotides corresponding to 338 amino acid residues. The protein molecular weight was estimated to be 36,294, which is in good agreement with the subunit molecular weight of the enzyme determined by polyacrylamide gel electrophoresis. The enzyme was overexpressed in recombinant Escherichia coli cells and purified to homogeneity by one hydrophobic column chromatography step. The predicted amino acid sequence showed no significant similarity to those of the currently known PLP-dependent enzymes but displayed 40 and 41% identity with those of the hypothetical GLY1 protein of Saccharomyces cerevisiae and the GLY1-like protein of Caenorhabditis elegans, respectively. Accordingly, L-allo-TA might represent a new type of PLP-dependent enzyme. To determine the PLP-binding site of the enzyme, all of the three conserved lysine residues of L-allo-TA were replaced by alanine by site-directed mutagenesis. The purified mutant enzymes, K51A and K224A, showed properties similar to those of the wild type, while the mutant enzyme K199A was catalytically inactive, with corresponding disappearance of the absorption maximum at 420 nm. Thus, Lys199 of L-allo-TA probably functions as an essential catalytic residue forming an internal Schiff base with PLP of the enzyme to catalyze the reversible aldol reaction.
Sigma-Aldrich
D-Threonine, ≥98% (TLC)