MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Adenomatous polyposis coli (APC)-independent regulation of beta-catenin degradation via a retinoid X receptor-mediated pathway.
Adenomatous polyposis coli (APC)-independent regulation of beta-catenin degradation via a retinoid X receptor-mediated pathway.
The Journal of biological chemistry (2003-05-29)
Jia-Hao Xiao, Corine Ghosn, Cory Hinchman, Chad Forbes, Jenny Wang, Nonna Snider, Allison Cordrey, Yi Zhao, Roshantha A S Chandraratna
ABSTRACT
Beta-catenin is a component of stable cell adherent complexes whereas its free form functions as a transcription factor that regulate genes involved in oncogenesis and metastasis. Free beta-catenin is eliminated by two adenomatous polyposis coli (APC)-dependent proteasomal degradation pathways regulated by glycogen synthase kinase 3beta (GSK3 beta) or p53-inducible Siah-1. Dysregulation of beta-catenin turnover consequent to mutations in critical genes of the APC-dependent pathways is implicated in cancers such as colorectal cancer. We have identified a novel retinoid X receptor (RXR)-mediated APC-independent pathway in the regulation of beta-catenin. In this proteasomal pathway, RXR agonists induce degradation of beta-catenin and RXR alpha and repress beta-catenin-mediated transcription. In vivo, beta-catenin interacts with RXR alpha in the absence of ligand, but RXR agonists enhanced the interaction. RXR agonist action was not impaired by GSK3 beta inhibitors or deletion of the GSK3 beta-targeted sequence from beta-catenin. In APC- and p53-mutated colorectal cancer cells, RXR agonists still inactivated endogenous beta-catenin via RXR alpha. Interestingly, deletion of the RXR alpha A/B region abolished ligand-induced beta-catenin degradation but not RXR alpha-mediated transactivation. RXR alpha-mediated inactivation of oncogenic beta-catenin paralleled a reduction in cell proliferation. These results suggest a potential role for RXR and its agonists in the regulation of beta-catenin turnover and related biological events.
Sigma-Aldrich
TOPflash (TCF Reporter Plasmid), Transfection grade T-cell factor reporter plasmid containing 2 sets (with the second set in the reverse orientation) of 3 copies of the TCF binding site (wild type) upstream of the Thymidine Kinase minimal promoter and Luciferase open reading frame.