MilliporeSigma
  • A small molecule very late antigen-4 antagonist can inhibit ovalbumin-induced lung inflammation.
A small molecule very late antigen-4 antagonist can inhibit ovalbumin-induced lung inflammation.
American journal of respiratory and critical care medicine (2003-02-06)
Gloria C Koo, Kashmira Shah, Gloria J F Ding, Jianying Xiao, Richard Wnek, George Doherty, Xin Chun Tong, R Blake Pepinsky, Ko-Chung Lin, William K Hagmann, Douglas Kawka, Irwin I Singer
ABSTRACT
A nonpeptidyl small molecule antagonist, compound A, to nonactivated very late antigen-4 (VLA4) was examined in lung inflammation induced by a single dose of ovalbumin challenge. Compound A presented a good pharmacokinetic property, when given intratracheally, and the blood cells from such pharmacokinetic study showed good receptor occupancy of the compound for approximately 8 hours. Compound A was then tested in an ovalbumin-induced airway inflammation model by intranasal or intravenous route of administration. There was a dose-dependent inhibition of eosinophilia in the bronchiolar lavage fluid, when compound A was given intranasally but not when it was given intravenously. For comparison, antibody to VLA4 and another compound, BIO1211, which reacts only with activated VLA4, were examined in this system. Immunohistochemical analyses of the lung tissue substantiated the findings in the bronchiolar lavage fluid. Specific staining of the major basic protein of eosinophils showed peribronchiolar infiltration of eosinophils. Some of these eosinophils were also positive for nitrotyrosine, suggesting activation of eosinophils in the lung interstitium. There was deposition of major basic protein and nitrotyrosine at the base of the perivascular endothelium, indicative of degranulation of eosinophils in the area. After intranasal treatment with compound A, eosinophils in the lungs and their activation products were substantially decreased, documenting its effectiveness in inhibiting lung inflammation.
Sigma-Aldrich
BIO 1211 trifluoroacetate salt, ≥97% (HPLC)