MilliporeSigma
Detection of NADPH-diaphorase activity in Paramecium primaurelia.
European journal of protistology (2006-10-31)
Andrea Amaroli, Marzia Ognibene, Francesca Trielli, Sonya Trombino, Carla Falugi, Maria Umberta Delmonte Corrado
ABSTRACT
Recently, we showed that Paramecium primaurelia synthesizes molecules functionally related to the cholinergic system and involved in modulating cell-cell interactions leading to the sexual process of conjugation. It is known that nitric oxide (NO) plays a role in regulating the release of transmitter molecules, such as acetylcholine, and that the NO biosynthetic enzyme, nitric oxide synthase (NOS), shows nicotinamide adenine dinucleotide phosphate-diaphorase (NADPH-d) activity. In this work, we detected the presence of NADPH-d activity in P. primaurelia. We characterized this activity histochemically by examining its specificity for beta-NADPH and alpha-NADH co-substrates, and sensitivity both to variations in chemico-physical parameters and to inhibitors of enzymes showing NADPH-d activity. Molecules immunologically related to NOS were recognized by the anti-rat brain NOS (bNOS) antibody. Moreover, bNOS immunoreactivity and NADPH-d activity sites were found to be co-localized. The non-denaturing electrophoresis, followed by exposure to beta-NADPH or alpha-NADH co-substrates, revealed the presence of a band of apparent molecular mass of about 124 kDa or a band of apparent molecular mass of about 175 kDa, respectively. In immunoblot experiments, the bNOS antibody recognized a single band of apparent molecular mass of about 123 kDa.
Sigma-Aldrich
α-Nicotinamide adenine dinucleotide, reduced disodium salt, ≥90%