MilliporeSigma
  • Heterologous expression, purification, immobilization and characterization of recombinant α-amylase AmyLa from Laceyella sp. DS3.
Heterologous expression, purification, immobilization and characterization of recombinant α-amylase AmyLa from Laceyella sp. DS3.
International journal of biological macromolecules (2019-04-07)
Ahmed K A El-Sayed, Mohamed I Abou-Dobara, Amira A El-Fallal, Noha F Omar
ABSTRACT
AmyLa α-amylase gene from Laceyella sp. DS3 was heterologously expressed in E. coli BL21. E. coli BL21 maximally expressed AmyLa after 4 h of adding 0.02 mM IPTG at 37 °C. The recombinant AmyLa α-amylase was purified 2.19-fold through gel filtration and ion exchange chromatography. We immobilized the purified recombinant AmyLa α-amylase on four carriers; chitosan had the best efficiency. The recombinant free and the immobilized AmyLa α-amylase showed optimum activity in the pH ranges of 6.0-7.0 and 4.0-7.0, respectively and possessed an optimum temperature of 55 °C. The free enzyme had activation energy, Km, and Vmax of 291.5 kJ, 1.5 mg/ml, and 6.06 mg/min, respectively. The immobilized enzyme had activation energy, Km, and Vmax of 309.74 kJ, 6.67 mg/ml, and 50 mg/min, respectively. The immobilized enzyme was calcium-independent and insensitive (relative to the free enzyme) to metals. It could also be reused for seven cycles.