MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Budding and Division of Giant Vesicles Linked to Phospholipid Production.
Budding and Division of Giant Vesicles Linked to Phospholipid Production.
Scientific reports (2019-01-19)
Juan M Castro, Hironori Sugiyama, Taro Toyota
ABSTRACT
The self-reproduction of supramolecular assemblies based on the synthesis and self-assembly of building blocks is a critical step towards the construction of chemical systems with autonomous, adaptive, and propagation properties. In this report, we demonstrate that giant vesicles can grow and produce daughter vesicles by synthesizing and incorporating phospholipids in situ from ad-hoc precursors. Our model involves acyl chain elongation via copper(I)-catalyzed azide-alkyne [3 + 2] cycloaddition reaction and the ensuing production of synthetic phospholipids to induce budding and division. In addition, the growth and budding of giant vesicles were compatible with the encapsulation and transfer of macromolecules as large as lambda phage DNA to the buds. This chemical system provides a useful model towards the implementation of cell-like compartments capable of propagation and transport of biological materials.