MilliporeSigma
  • Acetylation as a major determinant to microtubule-dependent autophagy: Relevance to Alzheimer's and Parkinson disease pathology.
Acetylation as a major determinant to microtubule-dependent autophagy: Relevance to Alzheimer's and Parkinson disease pathology.
Biochimica et biophysica acta. Molecular basis of disease (2018-12-21)
A R Esteves, A M Palma, R Gomes, D Santos, D F Silva, S M Cardoso
ABSTRACT
Protein post-translational modifications (PTMs) that potentiate protein aggregation have been implicated in several neurological disorders, including Alzheimer's (AD) and Parkinson's disease (PD). In fact, Tau and alpha-synuclein (ASYN) undergo several PTMs potentiating their aggregation and neurotoxicity. Recent data posits a role for acetylation in Tau and ASYN aggregation. Herein we aimed to clarify the role of Sirtuin-2 (SIRT2) and HDAC6 tubulin deacetylases as well as p300 acetyltransferase in AD and PD neurodegeneration. We used transmitochondrial cybrids that recapitulate pathogenic alterations observed in sporadic PD and AD patient brains and ASYN and Tau cellular models. We confirmed that Tau protein and ASYN are microtubules (MTs)-associated proteins (MAPs). Moreover, our results suggest that α-tubulin acetylation induced by SIRT2 inhibition is functionally associated with the improvement of MT dynamic determined by decreased Tau phosphorylation and by increased Tau/tubulin and ASYN/tubulin binding. Our data provide a strong evidence for a functional role of tubulin and MAPs acetylation on autophagic vesicular traffic and cargo clearance. Additionally, we showed that an accumulation of ASYN oligomers imbalance mitochondrial dynamics, which further compromise autophagy. We also demonstrated that an increase in Tau acetylation is associated with Tau phosphorylation. We found that p300, HDAC6 and SIRT2 influences Tau phosphorylation and autophagic flux in AD. In addition, we demonstrated that p300 and HDAC6 modulate Tau and Tubulin acetylation. Overall, our data disclose the role of Tau and ASYN modifications through acetylation in AD and PD pathology, respectively. Moreover, this study indicates that MTs can be a promising therapeutic target in the field of neurodegenerative disorders in which intracellular transport is altered.