MilliporeSigma
  • Sulindac-derived retinoid X receptor-α modulator attenuates atherosclerotic plaque progression and destabilization in ApoE-/- mice.
Sulindac-derived retinoid X receptor-α modulator attenuates atherosclerotic plaque progression and destabilization in ApoE-/- mice.
British journal of pharmacology (2019-04-04)
Linghong Shen, Zhe Sun, Peng Nie, Ruosen Yuan, Zhaohua Cai, Caizhe Wu, Liuhua Hu, Shuxuan Jin, Hu Zhou, Xiaokun Zhang, Ben He
ABSTRACT
Atherosclerosis is a chronic inflammatory disease, and retinoid X receptor-α (RXRα) is an intriguing anti-atherosclerosis target. This study investigated whether and how an RXRα modulator, K-80003, derived from a non-steroidal anti-inflammatory drug attenuates atherosclerotic plaque progression and destabilization. Our previously established ApoE-/- mouse model of carotid vulnerable plaque progression was treated with K-80003 or vehicle for 4 or 8 weeks. Samples of carotid arteries and serum were collected to determine atherosclerotic lesion size, histological features, expression of related proteins, and lipid profiles. In vitro studies were carried out in 7-ketocholesterol (7-KC)-stimulated macrophages treated with or without K-80003. K-80003 significantly reduced lesion size, plaque rupture, macrophage infiltration, and inflammatory cytokine levels. Plaque macrophages positive for nuclear p65 (RelA) NF-κB subunit were markedly reduced after K-80003 treatment. Also, K-80003 treatment inhibited 7-KC-induced p65 nuclear translocation, IκBα degradation, and transcription of NF-κB target genes. In addition, K-80003 inhibited NF-κB pathway mainly through the reduction of p62/sequestosome 1 (SQSTM1), probably due to promotion of autophagic flux by K-80003. Mechanistically, cytoplasmic localization of RXRα was associated with decreased autophagic flux. Increasing cytoplasmic RXRα expression by overexpression of RXRα/385 mutant decreased autophagic flux in RAW264.7 cells. Finally, K-80003 strongly inhibited 7-KC-induced RXRα cytoplasmic translocation. K-80003 suppressed atherosclerotic plaque progression and destabilization by promoting macrophage autophagic flux and consequently inhibited the p62/SQSTM1-mediated NF-κB proinflammatory pathway. Thus, targeting RXRα-mediated autophagy-inflammation axis by its noncanonical modulator may represent a promising strategy to treat atherosclerosis.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)