MilliporeSigma
  • Serum Aldosterone Is Related to Left Ventricular Geometry and Function in Young Adults with Never-Treated Primary Hypertension.
Serum Aldosterone Is Related to Left Ventricular Geometry and Function in Young Adults with Never-Treated Primary Hypertension.
Journal of clinical medicine (2019-07-20)
Seong-Mi Park, Mi-Na Kim, Sua Kim, Wan-Joo Shim
ABSTRACT
Although aldosterone has been demonstrated to induce left ventricular (LV) hypertrophy not only in primary aldosteronism but also in primary hypertension (HT), it can be affected by multiple factors, including age, and the effect of aldosterone on LV function is controversial. This study was to investigate the relationship of aldosterone to changes in LV geometry and function in young adults with never-treated HT. Seventy-five consecutive patients (age, 29.8 ± 6.3 years) with never-treated HT and 45 normal controls were enrolled. Echocardiographic values and LV global longitudinal strain (LVGLS) were obtained. Serum aldosterone concentration (SAC) and serum procollagen type III amino-terminal peptide (PIIINP) level were obtained in HT patients. HT patients had higher LV mass index, higher relative wall thickness (RWT), and worse LV function than normal controls. LVGLS and e' velocity were worse in HT patients with normal geometry than in normal controls. SAC was well correlated with LV mass index, RWT, e' velocity, LVGLS, and PIIINP (all p < 0.05). LV geometry pattern was most related to SAC among clinical parameters (p = 0.019). LVGLS was most related to LV geometry and diastolic blood pressure. In contrast, e' velocity was most related to PIIINP. Our findings may indicate that in young patients with never-treated HT, aldosterone significantly contributes to changes in LV geometry and functional impairment through its pro-hypertrophic and myocardial fibrosis effects beyond blood pressure.