MilliporeSigma
  • Cell migration capability of vascular endothelial growth factor into the root apex of a root canal model in vivo.
Cell migration capability of vascular endothelial growth factor into the root apex of a root canal model in vivo.
Journal of oral science (2018-06-01)
Takashi Matsuura, Kouji Sugimoto, Viviane K S Kawata-Matsuura, Kajiro Yanagiguchi, Shizuka Yamada, Yoshihiko Hayashi
ABSTRACT
Once a tooth develops deep caries and the dental pulp tissue is irreversibly infected, the infected dental pulp tissue should be removed, and filling material should be placed in the root canal. Endodontically treated teeth are prone to root fracture or periapical periodontitis; however, dental pulp tissue has the potential to prevent root fracture or periapical periodontitis. Therefore, dental pulp regeneration after pulpectomy may help prolong tooth life. In this study, a new method of dental pulp regeneration was developed. Vascular endothelial growth factor-adsorbed collagen gel was injected into the root canal of a prepared root canal model, placed into the dorsum of a rat, and cultured for 3 weeks. After retrieving the implant, histological analysis was performed. It was found that rat somatic cells were recruited into the root apex of the transplanted root canal model. These findings suggest a new potential technique for engineering dental pulp tissue.