Merck
  • Mechanism of Interleukin-4 Reducing Lipid Deposit by Regulating Hormone-Sensitive Lipase.
Mechanism of Interleukin-4 Reducing Lipid Deposit by Regulating Hormone-Sensitive Lipase.
Scientific reports (2019-08-21)
Ming-Yuh Shiau, Pei-Hua Chuang, Ching-Ping Yang, Chiao-Wan Hsiao, Shu-Wen Chang, Kai-Yun Chang, Tsung-Ming Liu, Huan-Wen Chen, Cheng-Chieh Chuang, Sheau-Yun Yuan, Yih-Hsin Chang
ABSTRACT
Accumulating evidence indicates that inflammation participates in the pathophysiological progress from insulin resistance, obesity, metabolic abnormalities, and type 2 diabetes mellitus. Our previous study reveals that interleukin-4 (IL-4) inhibits adipogenesis and promotes lipolysis to decrease lipid deposits by enhancing the activity of hormone sensitive lipase (HSL). The present study further dissects and characterizes the molecular mechanism of IL-4 in regulating HSL expression and lipolytic activity in the terminal differentiated 3T3-L1 mature adipocytes. Our results showed that IL-4 increased cAMP which then enhanced PKA activity and subsequent phosphorylation of HSL and perilipin. The phosphorylated HSL (p-HSL) translocated from cytoplasm to the surface of lipid droplets and exhibited lipolytic function. After being phosphorylated, p-perilipin also facilitated lipolysis through interacting with p-HSL. The in vitro findings were further verified by in vivo study in which IL-4 exhibited pro-lipolytic activity and enhanced HSL activity. In summary, the net outcome of IL-4 treatment is to reduce lipid storage by promoting lipolysis through enhancing HSL activity via cAMP/PKA pathway, the major route leading to lipolysis.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.