MilliporeSigma
Cascade biotransformation of estrogens by Isaria fumosorosea KCh J2.
Scientific reports (2019-07-26)
Ewa Kozłowska, Monika Dymarska, Edyta Kostrzewa-Susłow, Tomasz Janeczko
ABSTRACT
Estrone, estradiol, ethynylestradiol and estrone 3-methyl ether underwent a biotransformation process in the submerged culture of Isaria fumosorosea KCh J2. Estrone was transformed into seven metabolites, four of which were glycosylated. Estradiol was selectively glycosylated at C-3 and then transformed to D-ring lactone. Ethynylestradiol was coupled with methylglucoside and 6β-hydroxyderivative was obtained. Estrone 3-methyl ether was not transformed indicating that a free hydroxyl group at C-3 is necessary for glycosylation. Baeyer-Villiger oxidation combined with hydroxylation and glycosylation was observed. All glycosides obtained in this study are 3-O-β-methylglucosides.