MilliporeSigma
  • Targeting Dopamine D2, Adenosine A2A, and Glutamate mGlu5 Receptors to Reduce Repetitive Behaviors in Deer Mice.
Targeting Dopamine D2, Adenosine A2A, and Glutamate mGlu5 Receptors to Reduce Repetitive Behaviors in Deer Mice.
The Journal of pharmacology and experimental therapeutics (2019-02-13)
Mark H Lewis, Christopher T Primiani, Amber M Muehlmann
ABSTRACT
Repetitive behaviors are seemingly purposeless patterns of behavior that vary little in form and are characteristic of many neurodevelopmental, psychiatric, and neurologic disorders. Our work has identified an association between hypofunctioning of the indirect basal ganglia pathway and the expression of repetitive behavior in the deer mouse model. In this study, we targeted indirect pathway cells of the striatum with single drugs and drug combinations that bind to dopamine D2, adenosine A2A, and glutamate mGlu5 receptors. These receptors function both individually and as receptor heteromers. We found that only the triple drug cocktail (L-741,626+CGS21680+CDPPB) that was designed to increase striatal indirect basal ganglia pathway cell function reduced repetitive behavior in adult male deer mice. No single drug or double drug combinations were effective at selectively reducing repetitive behavior. We found this triple drug cocktail reduced repetitive behavior in both short-acting and long-acting formulations and was effective throughout 7 days of daily administration. Conversely, another triple drug cocktail (quinpirole+SCH58261+MTEP) that was designed to further reduce striatal indirect basal ganglia pathway cell function caused a significant increase in repetitive behavior. Significant and behaviorally selective effects on repetitive behavior were only achieved with the triple drug cocktails that included doses of L-741,626 and quinpirole that have off-target effects (e.g., dopamine D3 receptors). These data further a role for decreased indirect basal ganglia pathway activation in repetitive behavior and suggest that targeting these receptors and/or heteromeric complexes on the indirect pathway neurons of the striatum may offer pharmacotherapeutic benefit for individuals with repetitive behavior disorders.