MilliporeSigma
Concise Approach to Cyclohexyne and 1,2-Cyclohexadiene Precursors.
The Journal of organic chemistry (2019-03-07)
Jason V Chari, Francesca M Ippoliti, Neil K Garg
ABSTRACT
Silyl triflate precursors to cyclic alkynes and allenes serve as valuable synthetic building blocks. We report a concise and scalable synthetic approach to prepare the silyl triflate precursors to cyclohexyne and 1,2-cyclohexadiene. The strategy involves a retro-Brook rearrangement of an easily accessible cyclohexanone derivative, followed by triflation protocols. This simple, yet controlled, method should enable the further study of strained alkynes and allenes in chemical synthesis.