MilliporeSigma
  • Solution processable liquid metal nanodroplets by surface-initiated atom transfer radical polymerization.
Solution processable liquid metal nanodroplets by surface-initiated atom transfer radical polymerization.
Nature nanotechnology (2019-05-22)
Jiajun Yan, Mohammad H Malakooti, Zhao Lu, Zongyu Wang, Navid Kazem, Chengfeng Pan, Michael R Bockstaller, Carmel Majidi, Krzysztof Matyjaszewski
ABSTRACT
Eutectic gallium indium (EGaIn) is a liquid metal alloy at room temperature. EGaIn microdroplets can be incorporated into elastomers to fabricate highly stretchable, mechanically robust, soft multifunctional composites with high thermal stability1 and electrical conductivity2-4 that are suitable for applications in soft robotics and self-healing electronics5-7. However, the current methods of preparation rely on mechanical mixing, which may lead to irregularly shaped micrometre-sized droplets and an anisotropic distribution of properties8. Therefore, procedures for the stabilization of sub-micrometre-sized droplets of EGaIn and compatibilization in polymer matrices and solvents have attracted significant attention9-12. Here we report the synthesis of EGaIn nanodroplets stabilized by polymeric ligand encapsulation. We use a surface-initiated atom transfer radical polymerization initiator to covalently functionalize the oxide layer on the surface of the EGaIn nanodroplets13 with poly(methyl methacrylate) (PMMA), poly(n-butyl acrylate) (PBMA), poly(2-dimethylamino)ethyl methacrylate) (PDMAEMA) and poly(n-butyl acrylate-block-methyl methacrylate) (PBA-b-PMMA). These nanodroplets are stable in organic solvents, in water or in polymer matrices up to 50 wt% concentration, enabling direct solution-casting into flexible hybrid materials. The liquid metal can be recovered from dispersion by acid treatment. The nanodroplets show good mechanical, thermal and optical properties, with a remarkable suppression of crystallization and melting temperatures (down to -80 °C from 15 °C).