MilliporeSigma
  • Smad7 Promotes Healing of Radiotherapy-Induced Oral Mucositis without Compromising Oral Cancer Therapy in a Xenograft Mouse Model.
Smad7 Promotes Healing of Radiotherapy-Induced Oral Mucositis without Compromising Oral Cancer Therapy in a Xenograft Mouse Model.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-09-07)
Jingjing Luo, Li Bian, Melanie A Blevins, Dongyan Wang, Chao Liang, Danfeng Du, Fanglong Wu, Barry Holwerda, Rui Zhao, David Raben, Hongmei Zhou, Christian D Young, Xiao-Jing Wang
ABSTRACT
We previously reported preventive and therapeutic effects of Smad7, a multifunctional protein, on radiotherapy (RT)-induced mucositis in mice without promoting human oral cancer cell survival or migration in vitro. The current study aims to determine whether a Smad7-based biologic can treat existing oral mucositis during radiotherapy for oral cancer and whether this treatment compromises RT-induced cancer cell killing in neighboring oral cancer.Experimental Design: We transplanted human oral cancer cells into the tongues of mice and applied craniofacial irradiation to simultaneously kill tumor cells and induce oral mucositis, thus modeling RT and mucositis in oral cancer patients. We topically applied a recombinant human Smad7 protein fused with the cell-penetrating Tat tag (Tat-Smad7) to the oral mucosa of tumor-bearing mice post RT when oral mucositis began to develop. Topically applied Tat-Smad7 penetrated cells in both the oral mucosa and oral cancer, attenuating TGFβ and NF-κB signaling as well as inflammation at both sites. Tat-Smad7 treatment alleviated oral mucositis with reductions in DNA damage and apoptosis in keratinocytes, but increased keratinocyte proliferation compared with vehicle-treated mucositis lesions. In contrast, adjacent oral cancer exposed to Tat-Smad7 did not show alterations in proliferation or direct DNA damage, but showed increased oxidative stress damage and apoptosis compared with tumors treated with vehicle. Our results suggest that short-course Tat-Smad7 application to oral mucositis promotes its healing but does not compromise the cytotoxic effect of RT on oral cancer and has context-specific effects on oral mucosa versus oral cancer.