MilliporeSigma
  • Non-canonical activation of β-catenin by PRL-3 phosphatase in acute myeloid leukemia.
Non-canonical activation of β-catenin by PRL-3 phosphatase in acute myeloid leukemia.
Oncogene (2018-10-12)
Phyllis S Y Chong, Jianbiao Zhou, Jing-Yuan Chooi, Zit-Liang Chan, Sabrina Hui Min Toh, Tuan Zea Tan, Sheena Wee, Jayantha Gunaratne, Qi Zeng, Wee-Joo Chng
ABSTRACT
Aberrant activation of Wnt/β-catenin signaling pathway is essential for the development of AML; however, the mechanistic basis for this dysregulation is unclear. PRL-3 is an oncogenic phosphatase implicated in the development of LSCs. Here, we identified Leo1 as a direct and specific substrate of PRL-3. Serine-dephosphorylated form of Leo1 binds directly to β-catenin, promoting the nuclear accumulation of β-catenin and transactivation of TCF/LEF downstream target genes such as cyclin D1 and c-myc. Importantly, overexpression of PRL-3 in AML cells displayed enhanced sensitivity towards β-catenin inhibition in vitro and in vivo, suggesting that these cells are addicted to β-catenin signaling. Altogether, our study revealed a novel regulatory role of PRL-3 in the sustenance of aberrant β-catenin signaling in AML. PRL-3 may serve as a biomarker to select for the subset of AML patients who are likely to benefit from treatment with β-catenin inhibitors. Our study presents a new avenue of cancer inhibition driven by PRL-3 overexpression or β-catenin hyperactivation.