MilliporeSigma
  • Syntaphilin Is a Novel Biphasic Biomarker of Aggressive Prostate Cancer and a Metastasis Predictor.
Syntaphilin Is a Novel Biphasic Biomarker of Aggressive Prostate Cancer and a Metastasis Predictor.
The American journal of pathology (2019-05-14)
Michael J Hwang, Kelly G Bryant, Jae H Seo, Qin Liu, Peter A Humphrey, Mary Ann C Melnick, Dario C Altieri, Marie E Robert
ABSTRACT
Easily accessible biomarkers that may inform on the metastatic potential of localized prostate cancer are urgently needed. Herein, we show that syntaphilin (SNPH), a molecule originally identified as a negative regulator of mitochondrial dynamics in neurons, is abundantly expressed in prostate cancer. SNPH distribution in prostate cancer is spatially biphasic, with high expression at the invasive front, correlating with increased proliferative rates, as determined by Ki-67 labeling, and reduced levels in the central tumor bulk, which are further decreased in patients with distant metastases. Higher levels of SNPH are observed with increasing Gleason grade. Prostate tumors predominantly express a novel, extraneuronal isoform of SNPH that accumulates in mitochondria and maintains oxidative metabolism and tumor cell proliferation. These data suggest that SNPH is a novel marker of high Gleason grade prostate cancer, differentially expressed at the invasive front compared with the central tumor bulk, and is potentially down-regulated in metastatic disease. This biphasic pattern of expression may reflect a dual function of SNPH in controlling the balance between cell proliferation and invasion in tumors.