MilliporeSigma
  • Bladder drug mirabegron exacerbates atherosclerosis through activation of brown fat-mediated lipolysis.
Bladder drug mirabegron exacerbates atherosclerosis through activation of brown fat-mediated lipolysis.
Proceedings of the National Academy of Sciences of the United States of America (2019-05-16)
Wenhai Sui, Hongshi Li, Yunlong Yang, Xu Jing, Fei Xue, Jing Cheng, Mei Dong, Meng Zhang, Huazheng Pan, Yuguo Chen, Yunjian Zhang, Qingjun Zhou, Weiyun Shi, Xinsheng Wang, Han Zhang, Cheng Zhang, Yun Zhang, Yihai Cao
ABSTRACT
Mirabegron (Myrbetriq) is a β3-adrenoreceptor agonist approved for treating overactive bladder syndrome in human patients. This drug can activate brown adipose tissue (BAT) in adult humans and rodents through the β3-adrenoreceptor-mediated sympathetic activation. However, the effect of the mirabegron, approved by the US Food and Drug Administration, on atherosclerosis-related cardiovascular disease is unknown. Here, we show that the clinical dose of mirabegron-induced BAT activation and browning of white adipose tissue (WAT) exacerbate atherosclerotic plaque development. In apolipoprotein E-/- (ApoE-/-) and low-density lipoprotein (LDL) receptor-/- (Ldlr-/-) mice, oral administration of clinically relevant doses of mirabegron markedly accelerates atherosclerotic plaque growth and instability by a mechanism of increasing plasma levels of both LDL-cholesterol and very LDL-cholesterol remnants. Stimulation of atherosclerotic plaque development by mirabegron is dependent on thermogenesis-triggered lipolysis. Genetic deletion of the critical thermogenesis-dependent protein, uncoupling protein 1, completely abrogates the mirabegron-induced atherosclerosis. Together, our findings suggest that mirabegron may trigger cardiovascular and cerebrovascular diseases in patients who suffer from atherosclerosis.