Merck
  • Single-Cell Transcriptomics of Traced Epidermal and Hair Follicle Stem Cells Reveals Rapid Adaptations during Wound Healing.
Single-Cell Transcriptomics of Traced Epidermal and Hair Follicle Stem Cells Reveals Rapid Adaptations during Wound Healing.
Cell reports (2018-10-18)
Simon Joost, Tina Jacob, Xiaoyan Sun, Karl Annusver, Gioele La Manno, Inderpreet Sur, Maria Kasper
ABSTRACT
Epithelial tissues, such as the skin, rely on cellular plasticity of stem cells (SCs) from different niches to restore tissue function after injury. How these molecularly and functionally diverse SC populations respond to injury remains elusive. Here, we genetically labeled Lgr5- or Lgr6-expressing cells from the hair follicle bulge and interfollicular epidermis (IFE), respectively, and monitored their individual transcriptional adaptations during wound healing using single-cell transcriptomics. Both Lgr5 and Lgr6 progeny rapidly induced a genetic wound signature that, for Lgr5 progeny, included the remodeling of receptors to permit interactions with the wound environment, a property that Lgr6 progeny possessed even before wounding. When contributing to re-epithelialization, Lgr5 progeny gradually replaced their bulge identity with an IFE identity, and this process started already before Lgr5 progeny left the bulge. Altogether, this study reveals how different SCs respond and adapt to a new environment, potentially explaining cellular plasticity of many epithelial tissues.
Sigma-Aldrich
Tamoxifen, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Versican Antibody, a.a. 1360-1439 of mouse versican, Chemicon®, from rabbit