MilliporeSigma
  • Pentosan Polysulfate Demonstrates Anti-human T-Cell Leukemia Virus Type 1 Activities In Vitro and In Vivo.
Pentosan Polysulfate Demonstrates Anti-human T-Cell Leukemia Virus Type 1 Activities In Vitro and In Vivo.
Journal of virology (2019-06-07)
Guangyong Ma, Jun-Ichirou Yasunaga, Koichi Ohshima, Tadashi Matsumoto, Masao Matsuoka
ABSTRACT
Human T-cell leukemia virus type 1 (HTLV-1) infection causes T-cell leukemia and inflammatory diseases, most notably including HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP). The underlying mechanism for the pathogenesis of HAM/TSP remains unclear. According to a recent clinical trial, a humanized antibody that targets CCR4+ cells ameliorates inflammation by reducing the number of infected cells in the central nervous system; this result suggests that the transmigration of HTLV-1-infected cells plays a crucial role in HAM/TSP. Partly due to the blood-brain barrier, current treatments for HAM/TSP are mostly palliative. Pentosan polysulfate (PPS), a semisynthetic glycosaminoglycan, has recently been used to treat HAM/TSP and was found to alleviate the symptoms. In this study, we investigated the effect of PPS on HTLV-1-infected cells and provide evidence for its efficacy in HAM/TSP. PPS was cytotoxic to certain HTLV-1-infected cells and significantly suppressed HTLV-1 virion production. PPS also efficiently inhibited HTLV-1 cell-cell transmission in T cells. In addition, PPS blocked HTLV-1 infection of primary endothelial cells (human umbilical vascular endothelial cells) and suppressed the subsequent induction of proinflammatory cytokine expression. Furthermore, PPS was found to inhibit the adhesion and transmigration of HTLV-1-infected cells. We also confirmed the anti-HTLV-1 effect of PPS in vivo using two mouse models. PPS blocked HTLV-1 infection in a mouse model with peripheral blood mononuclear cell (PBMC)-humanized NOD-scid IL2Rgammanull (huPBMC NSG) mice. PPS was also found to suppress the development of dermatitis and lung damage in HTLV-1 bZIP factor (HBZ)-transgenic (HBZ-Tg) mice, an HTLV-1 transgenic mouse model in which the mice develop systemic inflammation.IMPORTANCE HTLV-1 is the first human retrovirus to have been identified and is endemic in certain areas worldwide. HTLV-1 infection leads to the development of an inflammatory disease called HAM/TSP, a myelopathy characterized by slowly progressive spastic paraparesis. There have been no effective therapeutics available for HAM/TSP, but recently, a semisynthetic glycosaminoglycan, named pentosan polysulfate (PPS), has been found to alleviate the symptoms of HAM/TSP. Here we conducted a comprehensive study on the effect of PPS both in vitro and in vivo PPS demonstrated anti-HTLV-1 potential in infected cell lines, as shown by its suppressive effects on HTLV-1 replication and transmission and on the transmigration of infected T cells. Moreover, results obtained from two HTLV-1 mouse models demonstrate that PPS inhibits HTLV-1 infection and inflammation development in vivo Our work offers insights into the treatment of HAM/TSP by PPS and also suggests its possible use for treating other HTLV-1-induced inflammatory diseases.