MilliporeSigma
  • Thymoquinone reduces intracytoplasmic oxidative stress and improves epigenetic modification in polycystic ovary syndrome mice oocytes, during in-vitro maturation.
Thymoquinone reduces intracytoplasmic oxidative stress and improves epigenetic modification in polycystic ovary syndrome mice oocytes, during in-vitro maturation.
Molecular reproduction and development (2019-06-19)
Fatemeh Eini, Arash Bidadkosh, Hamid Nazarian, Abbas Piryaei, Marefat Ghaffari Novin, Khojasteh Joharchi
ABSTRACT
Although in-vitro maturation (IVM) of oocytes has been presented as an alternative treatment to traditional stimulated in-vitro fertilization, the culture condition can be improved by natural antioxidants. Thus, we investigated the protective effect of Thymoquinone (TQ) during IVM in the polycystic ovary syndrome (PCOS) mice model. The induction of PCOS was made by dehydroepiandrosterone via subcutaneous injection, in prepubertal female B6D2F1-mice. After 21 days later, germinal vesicle (GV)-stage-oocytes were extracted and incubated in IVM media containing 0, 1.0, 10.0, and 100.0 μM of TQ. To assess fertilization and blastulation rates, after 22-24 hr, the treated oocytes were fertilized in-vitro with epididymal spermatozoa. Some other oocytes were evaluated for maturation, epigenetic, and oxidative stress markers. Similarly, the mRNA expression of epigenetic enzymes genes (Dnmt1 and Hdac1), three maternally derived genes (Mapk, CyclinB, and Cdk1) and apoptosis-related genes (Bax and Bcl2) were assessed. Our results showed that the maturation, fertilization, and blastulation rates were significantly higher in the 10.0 μM TQ-treated group compared with the untreated group and likewise with in-vivo matured oocytes. The Bax expression was reduced in 10.0 μM TQ matured oocytes, but Bcl2, Dnmt1, Hdac1, Cdk1, and Mapk were upregulated in this group compared to other groups. Furthermore, dimethylation of histone-3 at lysine-9 (H3K9m2) and DNA methylation were significantly increased whereas H4K12 acetylation (H4K12ac) was decreased in the 10.0 μM TQ-treated group in comparison with control and in-vivo matured oocytes. Therefore, our results are suggesting that 10.0 μM TQ may enhance the developmental competence of PCOS oocytes via the modulation of oxidative stress and epigenetic alterations.