MilliporeSigma
Brain-transportable dipeptides across the blood-brain barrier in mice.
Scientific reports (2019-04-10)
Mitsuru Tanaka, Shinya Dohgu, Genki Komabayashi, Hayato Kiyohara, Fuyuko Takata, Yasufumi Kataoka, Takashi Nirasawa, Motohiro Maebuchi, Toshiro Matsui
ABSTRACT
Apart from nutrients required for the brain, there has been no report that naturally occurring peptides can cross the blood-brain barrier (BBB). The aim of this study was to identify the BBB-transportable peptides using in situ mouse perfusion experiments. Based on the structural features of Gly-N-methylated Gly (Gly-Sar), a reported BBB-transportable compound, 18 dipeptides were synthesized, and were perfused in the mouse brain for two minutes. Among the synthesized dipeptides, Gly-Sar, Gly-Pro, and Tyr-Pro were transported across the BBB with Ki values of 7.60 ± 1.29, 3.49 ± 0.66, and 3.53 ± 0.74 µL/g·min, respectively, and accumulated in the mouse brain parenchyma. Additionally, using MALDI-MS/MS imaging analysis of Tyr-Pro-perfused brain, we provide evidence for Tyr-Pro accumulation in the hippocampus, hypothalamus, striatum, cerebral cortex, and cerebellum of mouse brain.