MilliporeSigma
  • Oleanane-type glycosides from the roots of Weigela florida "rumba" and evaluation of their antibody recognition.
Oleanane-type glycosides from the roots of Weigela florida "rumba" and evaluation of their antibody recognition.
Fitoterapia (2018-05-04)
Anne-Sophie Champy-Tixier, Anne-Claire Mitaine-Offer, Feliciana Real Fernández, Tomofumi Miyamoto, Chiaki Tanaka, Anna-Maria Papini, Marie-Aleth Lacaille-Dubois
ABSTRACT
Three triterpene glycosides were isolated from the roots of Weigela florida "rumba" (Bunge) A. DC.: two previously undescribed 3-O-β-d-xylopyranosyl-(1→2)-[β-d-xylopyranosyl-(1→4)]-β-d-xylopyranosyl-(1→4)-β-d-xylopyranosyl-(1→3)-α-L-rhamnopyranosyl-(1→2)-α-l-arabinopyranosyloleanolic acid (1) and 3-O-β-d-xylopyranosyl-(1→2)-[β-d-glucopyranosyl-(1→4)]-β-d-xylopyranosyl-(1→4)-β-d-xylopyranosyl-(1→3)-α-l-rhamnopyranosyl-(1→2)-α-L-arabinopyranosyloleanolic acid (2), and one isolated for the first time from a natural source 3-O-β-d-xylopyranosyl-(1→3)-α-l-rhamnopyranosyl-(1→2)-α-l-arabinopyranosyloleanolic acid (3). Their structures were elucidated mainly by 2D NMR spectroscopic analysis (COSY, TOCSY, NOESY, HSQC, HMBC) and mass spectrometry. Compounds 2 and 3 were further evaluated as antigens in enzyme-linked immunosorbent assay (ELISA) to recognize IgM antibodies in multiple sclerosis (MS) patients' sera.