MilliporeSigma
  • Stearoyl-Chitosan Coated Nanoparticles Obtained by Microemulsion Cold Dilution Technique.
Stearoyl-Chitosan Coated Nanoparticles Obtained by Microemulsion Cold Dilution Technique.
International journal of molecular sciences (2018-12-06)
Daniela Chirio, Elena Peira, Simona Sapino, Chiara Dianzani, Alessandro Barge, Elisabetta Muntoni, Silvia Morel, Marina Gallarate
ABSTRACT
Chitosan is an excipient which has been studied thoroughly in research works thanks to its positive characteristics such as muco-adhesiveness and ability to open epithelial-tight-junctions. In this article, lipophilic stearoyl chitosan (ST-CS) was synthetized in order to anchor this polymer to lipid nanoparticles and prepare ST-CS-coated nanoparticles (ST-CS-NP) using the microemulsion cold dilution technique. Curcumin (CURC) was used as model drug. CURC-ST-CS-NP were characterized by dimensional analysis, zeta potential, drug entrapment, drug release; tested in vitro on Human Umbilical Vein Endothelial Cell (HUVEC) cells to study its cytotoxicity and on human pancreatic cancer cells (PANC-1) to determine inhibition ability; tested in rats to determine CURC blood profiles and biodistribution. CURC-ST-CS-NP had mean diameters in the range 200⁻400 nm and CURC entrapment up to 73%. These systems did not show cytotoxicity on HUVEC cells at all tested dilutions and revealed to be more effective than free CURC solution on PANC-1 cells at 5 and 10 µM CURC. Blood profile studies evidenced as CURC entrapment in NP prolonged the permanence of drug in the systemic circulation compared to CURC solution due to a certain stealth property of NP, probably attributable to hydrophilic chitosan coating. Biodistribution studies showed a smaller CURC concentration in RES organs when CURC-ST-CS-NP were administered.