MilliporeSigma
  • p62/SQSTM1 Fuels Melanoma Progression by Opposing mRNA Decay of a Selective Set of Pro-metastatic Factors.
p62/SQSTM1 Fuels Melanoma Progression by Opposing mRNA Decay of a Selective Set of Pro-metastatic Factors.
Cancer cell (2018-12-26)
Panagiotis Karras, Erica Riveiro-Falkenbach, Estela Cañón, Cristina Tejedo, Tonantzin G Calvo, Raúl Martínez-Herranz, Direna Alonso-Curbelo, Metehan Cifdaloz, Eva Perez-Guijarro, Gonzalo Gómez-López, Pilar Ximenez-Embun, Javier Muñoz, Diego Megias, David Olmeda, Jorge Moscat, Pablo L Ortiz-Romero, Jose L Rodríguez-Peralto, María S Soengas
ABSTRACT
Modulators of mRNA stability are not well understood in melanoma, an aggressive tumor with complex changes in the transcriptome. Here we report the ability of p62/SQSTM1 to extend mRNA half-life of a spectrum of pro-metastatic factors. These include FERMT2 and other transcripts with no previous links to melanoma. Transcriptomic, proteomic, and interactomic analyses, combined with validation in clinical biopsies and mouse models, identified a selected set of RNA-binding proteins (RBPs) recruited by p62, with IGF2BP1 as a key partner. This p62-RBP interaction distinguishes melanoma from other tumors where p62 controls autophagy or oxidative stress. The relevance of these data is emphasized by follow-up analyses of patient prognosis revealing p62 and FERMT2 as adverse determinants of disease-free survival.