MilliporeSigma
  • Inhibition of microRNA-429 attenuates oxygen-glucose deprivation/reoxygenation-induced neuronal injury by promoting expression of GATA-binding protein 4.
Inhibition of microRNA-429 attenuates oxygen-glucose deprivation/reoxygenation-induced neuronal injury by promoting expression of GATA-binding protein 4.
Neuroreport (2018-04-07)
Jie Xiao, Ranran Kong, Jingye Hu
ABSTRACT
MicroRNAs (miRNAs) have been documented as critical regulators in ischemia/reperfusion-induced neuronal death. A better understanding of miRNA-mediated molecular mechanisms in ischemia/reperfusion-induced neuronal death may provide therapeutic targets for cerebral ischemia/reperfusion injury. A growing body of evidence suggests that miR-429 is a apoptosis-related miRNA that is also induced by hypoxia. However, whether miR-429 is involved in regulating neuronal apoptosis during cerebral ischemia/reperfusion injury remains unclear. In this study, the effect of miR-429 on oxygen-glucose deprivation and reoxygenation (OGD/R)-induced neuronal injury was investigated in vitro. The results showed that miR-429 expression levels were upregulated in cultured neurons with OGD/R treatment. The downregulation of miR-429 significantly alleviated OGD/R-induced neuronal injury, whereas upregulation of miR-429 aggravated it. Bioinformatic analysis showed that miR-429 could directly target the 3'-untranslated region of GATA-binding protein 4 (GATA4), which was verified by dual-luciferase reporter assay. Moreover, we found that miR-429 negatively regulated GATA4 expression. Overexpression of GATA4 also significantly alleviated OGD/R-induced neuronal injury. However, knockdown of GATA4 partially reversed the protective effect induced by miR-429 downregulation. Overall, our data showed that downregulation of miR-429 protected neurons against OGD/R-induced injury by promoting GATA4 and suggested a potential therapeutic target for the treatment of cerebral ischemia/reperfusion injury.