MilliporeSigma
  • A positively charged tight UF membrane and its properties for removing trace metal cations via electrostatic repulsion mechanism.
A positively charged tight UF membrane and its properties for removing trace metal cations via electrostatic repulsion mechanism.
Journal of hazardous materials (2019-03-27)
Ming-Yong Zhou, Peng Zhang, Li-Feng Fang, Bao-Ku Zhu, Jian-Li Wang, Jiang-Hua Chen, Heba Abdallah
ABSTRACT
The development of highly efficient membranes technology using low-pressure driven filtration process, is one of the principal challenges in the wastewater treatment field, especially those aimed at the removal of trace heavy metals. In this work, a novel positively charged tight ultrafiltration (PCTUF) membrane was developed to remove heavy metal cations (Mn2+, Co2+, Ni2+, Zn2+ and Cd2+) from contaminated waters via electrostatic repulsion mechanism. The PCTUF membrane was fabricated from a new polymer with poly (vinyl chloride co dimethylaminoethyl methacrylate), P (VC-co-DMA) via a nonsolvent induce phase separation (NIPS) process and following facile surface quaternization. The quaternization conditions, the pore structures and chemical properties of the membranes were investigated in detail. The optimally quaternized membrane possessed a positively charged surface and 3.27 nm charged channel with the water permeability of 84 L m-2 h-1 bar-1. The rejections of heavy metal cations surpassed 95% for feed solutions containing 10 ppm heavy metal. Moreover, the influences of feed concentrations and the operating condition with pressure and pH on the membrane performances were also investigated. The results revealed that the prepared PCTUF membrane with its high perm-selectivity performance provides a worthy reference for highly efficient removal of heavy metal cations.