MilliporeSigma
  • A Multiphase Protocol for Selective Hydrogenation and Reductive Amination of Levulinic Acid with Integrated Catalyst Recovery.
A Multiphase Protocol for Selective Hydrogenation and Reductive Amination of Levulinic Acid with Integrated Catalyst Recovery.
ChemSusChem (2019-04-17)
Alessandro Bellè, Tommaso Tabanelli, Giulia Fiorani, Alvise Perosa, Fabrizio Cavani, Maurizio Selva
ABSTRACT
At 60-150 °C and 15-35 bar H2 , two model reactions of levulinic acid (LA), hydrogenation and reductive amination with cyclohexylamine, were explored in a multiphase system composed of an aqueous solution of reactants, a hydrocarbon, and commercial 5 % Ru/C as a heterogeneous catalyst. By tuning the relative volume of the immiscible water/hydrocarbon phases and the concentration of the aqueous solution, a quantitative conversion of LA was achieved with formation of γ-valerolactone or N-(cyclohexylmethyl)pyrrolidone in >95 and 88 % selectivity, respectively. Additionally, the catalyst could be segregated in the hydrocarbon phase and recycled in an effective semi-continuous protocol. Under such conditions, formic acid additive affected the reactivity of LA through a competitive adsorption on the catalyst surface. This effect was crucial to improve selectivity for the reductive amination process. The comparison of 5 % Ru/C with a series of carbon supports demonstrated that the segregation phenomenon in the hydrocarbon phase, never previously reported, was pH-dependent and effective for samples displaying a moderate surface acidity.