MilliporeSigma
  • Determination of Fumonisin B1 in maize using molecularly imprinted polymer nanoparticles-based assay.
Determination of Fumonisin B1 in maize using molecularly imprinted polymer nanoparticles-based assay.
Food chemistry (2019-07-02)
Hasim Munawar, Abeer H M Safaryan, Annalisa De Girolamo, Alvaro Garcia-Cruz, Pedro Marote, Kal Karim, Vincenzo Lippolis, Michelangelo Pascale, Sergey A Piletsky
ABSTRACT
Fumonisin B1 (FB1) is a carcinogenic mycotoxin produced by Fusarium species contaminating maize. At present, fumonisin determination is performed using costly and demanding chromatography techniques or immunoassays. Recently, a molecularly imprinted polymer nanoparticles (nanoMIPs) - based assay (MINA) has been developed for FB1 detection. Herein, we have applied MINA for the determination of FB1 in naturally contaminated maize samples and results were compared with those obtained with ELISA and a reference HPLC method (AOAC No. 2001.04). The nanoMIPs as a recognition element mimicking antibodies used in ELISA were produced by solid phase synthesis and used in MINA for FB1 determination in 53 maize samples. As a result, 18 maize samples were contaminated with FB1 at levels higher than 0.25 mg/kg. Fumonisin concentrations from samples measured by MINA were well correlated with those using ELISA and HPLC. Therefore, MINA could be used as an alternative technique for FB1 determination in maize.