MilliporeSigma
  • Histone H3K27 Demethylase Negatively Controls the Memory Formation of Antigen-Stimulated CD8+ T Cells.
Histone H3K27 Demethylase Negatively Controls the Memory Formation of Antigen-Stimulated CD8+ T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2019-01-11)
Takeshi Yamada, Shogo Nabe, Koji Toriyama, Junpei Suzuki, Kazuki Inoue, Yuuki Imai, Atsushi Shiraishi, Katsuto Takenaka, Masaki Yasukawa, Masakatsu Yamashita
ABSTRACT
Although the methylation status of histone H3K27 plays a critical role in CD4+ T cell differentiation and its function, the role of Utx histone H3K27 demethylase in the CD8+ T cell-dependent immune response remains unclear. We therefore generated T cell-specific Utxflox/flox Cd4-Cre Tg (Utx KO) mice to determine the role of Utx in CD8+ T cells. Wild-type (WT) and Utx KO mice were infected with Listeria monocytogenes expressing OVA to analyze the immune response of Ag-specific CD8+ T cells. There was no significant difference in the number of Ag-specific CD8+ T cells upon primary infection between WT and Utx KO mice. However, Utx deficiency resulted in more Ag-specific CD8+ T cells upon secondary infection. Adoptive transfer of Utx KO CD8+ T cells resulted in a larger number of memory cells in the primary response than in WT. We observed a decreased gene expression of effector-associated transcription factors, including Prdm1 encoding Blimp1, in Utx KO CD8+ T cells. We confirmed that the trimethylation level of histone H3K27 in the Prdm1 gene loci in the Utx KO cells was higher than in the WT cells. The treatment of CD8+ T cells with Utx-cofactor α-ketoglutarate hampered the memory formation, whereas Utx inhibitor GSK-J4 enhanced the memory formation in WT CD8+ T cells. These data suggest that Utx negatively controls the memory formation of Ag-stimulated CD8+ T cells by epigenetically regulating the gene expression. Based on these findings, we identified a critical link between Utx and the differentiation of Ag-stimulated CD8+ T cells.