MilliporeSigma
  • Macrolides Inhibit Capsule Formation of Highly Virulent Cryptococcus gattii and Promote Innate Immune Susceptibility.
Macrolides Inhibit Capsule Formation of Highly Virulent Cryptococcus gattii and Promote Innate Immune Susceptibility.
Antimicrobial agents and chemotherapy (2019-04-03)
Shigeki Nakamura, Yurika Ikeda-Dantsuji, Lianjin Jin, Yoshitsugu Higashi, Masahiro Abe, Tatsuya Inukai, Minoru Nagi, Makoto Urai, Yoshitsugu Miyazaki
ABSTRACT
Cryptococcus gattii is a fungal pathogen, endemic in tropical and subtropical regions, the west coast of Canada, and the United States, that causes a potentially fatal infection in otherwise healthy individuals. Because the cryptococcal polysaccharide capsule is a leading virulence factor due to its resistance against innate immunity, the inhibition of capsule formation may be a promising new therapeutic strategy for C. gattii Macrolides have numerous nonantibiotic effects, including immunomodulation of mammalian cells and suppression of bacterial (but not fungal) pathogenicity. Thus, we hypothesized that a macrolide would inhibit cryptococcal capsule formation and improve the host immune response. Coincubation with clarithromycin (CAM) and azithromycin significantly reduced the capsule thickness and the amount of capsular polysaccharide of both C. gattii and C. neoformans CAM-treated C. gattii cells were significantly more susceptible to H2O2 oxidative stress and opsonophagocytic killing by murine neutrophils. In addition, more C. gattii cells were phagocytosed by murine macrophages, resulting in increased production of tumor necrosis factor alpha (TNF-α) by CAM exposure. After CAM exposure, dephosphorylation of Hog1, one of the mitogen-activated protein kinase (MAPK) signaling pathways of Cryptococcus, was observed in Western blot analysis. In addition, CAM exposure significantly reduced the mRNA expression of LAC1 and LAC2 (such mRNA expression is associated with cell wall integrity and melanin production). These results suggest that CAM may aid in inhibiting capsular formation via the MAPK signaling pathway and by suppressing virulent genes; thus, it may be a useful adjunctive agent for treatment of refractory C. gattii infection.