MilliporeSigma
  • Selective MT(2) melatonin receptor antagonist blocks melatonin-induced antinociception in rats.
Selective MT(2) melatonin receptor antagonist blocks melatonin-induced antinociception in rats.
Neuroscience letters (2000-03-16)
C X Yu, C B Zhu, S F Xu, X D Cao, G C Wu
ABSTRACT
The present study was undertaken to assess the effects of intracerebroventricular (i.c.v.) luzindole (a selective MT(2) melatonin receptor antagonist) and prazosin (a selective MT(3) melatonin receptor antagonist) on melatonin-induced antinociception, so as to clarify which of melatonin receptor subtypes within the central nervous system (CNS) was mediating antinociception. The pain threshold of rats was measured by the hot water (50 degrees C) tail-flick test. It was found that intraperitoneal (i.p.) melatonin (30, 60, 120 mg/kg) resulted in a dose-dependent antinociception. Luzindole (50, 100 microgram) administered intracerebroventricularly antagonized significantly the antinociceptive effect induced by i.p. melatonin (120 mg/kg), whereas prazosin (50 microgram) did not. Neither luzindole (100 microgram, i.c.v.) nor prazosin (50 microgram, i.c.v.) affected the nociceptive threshold when given alone. The results suggest that melatonin-induced antinociception is mediated through the MT(2) melatonin receptor subtype within the CNS.
Sigma-Aldrich
Luzindole, ≥90%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00