Merck
  • Evaluation of the immune humoral response of Brazilian patients with Rubinstein-Taybi syndrome.
Evaluation of the immune humoral response of Brazilian patients with Rubinstein-Taybi syndrome.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2010-11-19)
L C Torres, S M M Sugayama, C Arslanian, M M Sales, M Carneiro-Sampaio
ABSTRACT
Rubinstein-Taybi syndrome (RTS) is a rare developmental disorder characterized by craniofacial dysmorphisms, broad thumbs and toes, mental and growth deficiency, and recurrent respiratory infections. RTS has been associated with CREBBP gene mutations, but EP300 gene mutations have recently been reported in 6 individuals. In the present study, the humoral immune response in 16 RTS patients with recurrent respiratory infections of possible bacterial etiology was evaluated. No significant differences between patients and 16 healthy controls were detected to explain the high susceptibility to respiratory infections: normal or elevated serum immunoglobulin levels, normal salivary IgA levels, and a good antibody response to both polysaccharide and protein antigens were observed. However, most patients presented high serum IgM levels, a high number of total B cell and B subsets, and also high percentiles of apoptosis, suggesting that they could present B dysregulation. The CREBBP/p300 family gene is extremely important for B-cell regulation, and RTS may represent an interesting human model for studying the molecular mechanisms involved in B-cell development.
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution