MilliporeSigma
  • PTX3 Modulates Neovascularization and Immune Inflammatory Infiltrate in a Murine Model of Fibrosarcoma.
PTX3 Modulates Neovascularization and Immune Inflammatory Infiltrate in a Murine Model of Fibrosarcoma.
International journal of molecular sciences (2019-09-20)
Tiziana Annese, Roberto Ronca, Roberto Tamma, Arianna Giacomini, Simona Ruggieri, Elisabetta Grillo, Marco Presta, Domenico Ribatti
ABSTRACT
Fibrosarcoma is an aggressive subtype of soft tissue sarcoma categorized in infantile/congenital-type and adult-type. Fibrosarcoma cells and its surrounding immune inflammatory infiltrates overexpress or induce the expression of fibroblast growth factor-2 (FGF-2) that have a crucial role in tumor progression and angiogenesis. The inflammation-associated long pentraxin 3 (PTX3) was found to reduce FGF-2-mediated angiogenesis, but its role on fibrosarcoma immune inflammatory infiltrate is still unknown. In this study, we have evaluated the PTX3 activity on immune infiltrating mast cells, macrophages and T-lymphocytes by immunohistochemistry on murine MC-TGS17-51 fibrosarcoma cells and on transgenic TgN(Tie2-hPTX3) mouse. In these fibrosarcoma models we found a reduced neovascularization and a significant decrease of inflammatory infiltrate. Indeed, we show that PTX3 reduces the level of complement 3 (C3) deposition reducing fibrosarcoma progression. In conclusion, we hypothesize that targeting fibrosarcoma microenvironment by FGF/FGFR inhibitors may improve treatment outcome.
Sigma-Aldrich
Hematoxylin solution according to Mayer