MilliporeSigma
  • The Effect of the ortho Nitro Group in the Solvolysis of Benzyl and Benzoyl Halides.
The Effect of the ortho Nitro Group in the Solvolysis of Benzyl and Benzoyl Halides.
International journal of molecular sciences (2019-08-21)
Kyoung-Ho Park, Chan Joo Rhu, Jin Burm Kyong, Dennis N Kevill
ABSTRACT
A kinetic study was carried out on the solvolysis of o-nitrobenzyl bromide (o-isomer, 1) and p-nitrobenzyl bromide (p-isomer, 3), and o-nitrobenzoyl chloride (o-isomer, 2) in a wide range of solvents under various temperatures. In all of the solvents without aqueous fluoroalcohol, the reactions of 1 were solvolyzed at a similar rate to those observed for 3, and the reaction rates of 2 were about ten times slower than those of the previously studied p-nitrobenzoyl chloride (p-isomer, 4). For solvolysis in aqueous fluoroalcohol, the reactivity of 2 was kinetically more reactive than 4. The l/m values of the extended Grunwald-Winstein (G-W) equation for solvolysis of 1 and 2 in solvents without fluoroalcohol content are all significantly larger than unity while those in all the fluoroalcohol solvents are less than unity. The role of the ortho-nitro group as an intramolecular nucleophilic assistant (internal nucleophile) in the solvolytic reaction of 1 and 2 was discussed. The results are also compared with those reported earlier for o-carbomethoxybenzyl bromide (5) and o-nitrobenzyl p-toluenesulfonate (7). From the product studies and the activation parameters for solvolyses of 1 and 2 in several organic hydroxylic solvents, mechanistic conclusions are drawn.
Sigma-Aldrich
2-Nitrobenzyl bromide, 98%