MilliporeSigma
  • Benzyl butyl phthalate (BBP) triggers the migration and invasion of hemangioma cells via upregulation of Zeb1.
Benzyl butyl phthalate (BBP) triggers the migration and invasion of hemangioma cells via upregulation of Zeb1.
Toxicology in vitro : an international journal published in association with BIBRA (2019-06-19)
Shuo Cui, Linlei Wang, Huafei Zhao, Fei Lu, Weixin Wang, Zhengjiang Yuan
ABSTRACT
Hemangioma (HA) are tumors formed by hyper-proliferation of vascular endothelial cells. As a potential endocrine disrupting chemical (EDC), benzyl butyl phthalate (BBP) can mimic estrogen to disturb the estrogenic signals. Our present study investigated the potential roles of phthalates on the progression of HA and found that 100 nM BBP can significantly trigger the migration and invasion of HA cells, which was evidenced by the results that BBP can induce the expression of matrix metalloproteinase (MMPs) and vimentin. Further, BBP can increase the expression of Zeb1, one powerful transcription factor for cell migration and invasion. Targeted inhibition of Zeb1 blocked BBP induced cell migration. Mechanistically, BBP can increase the mRNA stability of Zeb1 via suppression of miR-655. Further, BBP can enhance the protein stability of Zeb1 via upregulation of ataxia telangiectasia mutated (ATM). Collectively, our present study revealed that BBP can trigger the migration and invasion of HA cells via upregulation of Zeb1.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%