MilliporeSigma
  • Effect of methylation of adenine N6 on kink turn structure depends on location.
Effect of methylation of adenine N6 on kink turn structure depends on location.
RNA biology (2019-06-27)
Saira Ashraf, Lin Huang, David M J Lilley
ABSTRACT
N6-methyladenine is the most common covalent modification in cellular RNA species, with demonstrated functional consequences. At the molecular level this methylation could alter local RNA structure, and/or modulate the binding of specific proteins. We have previously shown that trans-Hoogsteen-sugar (sheared) A:G base pairs can be completely disrupted by methylation, and that this occurs in a sub-set ofD/D k-turn structures. In this work we have investigated to what extent sequence context affects the severity with which inclusion of N6-methyladenine into different A:G base pairs of a standard k-turn affects RNA folding and L7Ae protein binding. We find that local sequence has a major influence, ranging from complete absence of folding and protein binding to a relatively mild effect. We have determined the crystal structure of one of these species both free and protein-bound, showing the environment of the methyl group and the way the modification is accommodated into the k-turn structure.
Sigma-Aldrich
Triethylamine trihydrofluoride, 97%